Assay Details
| Gene Symbol: Ankhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175375.3 | NP_780584.2 | 21 - 22 | 4087 | 89 | |
| GenBank mRNA | AK017018.1 | - | 11 - 12 | 1426 | 89 | |
| AK029322.1 | - | 14 - 15 | 2502 | 89 | ||
| AK042754.1 | - | 8 - 9 | 1062 | 89 | ||
| AK131166.1 | - | 21 - 22 | 3949 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankhd1
Genomic Map