Assay Details
| Gene Symbol: Ankhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175375.3 | NP_780584.2 | 6 - 7 | 1285 | 106 | |
| GenBank mRNA | AK131166.1 | - | 6 - 7 | 1148 | 106 | |
| CF742822.1 | - | 1 - 2 | 157 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankhd1
Genomic Map