Assay Details
| Gene Symbol: Tspan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028841.3 | NP_083117.2 | 2 - 3 | 327 | 65 | |
| GenBank mRNA | AK008761.1 | - | 2 - 3 | 272 | 65 | |
| AK166272.1 | - | 2 - 3 | 330 | 65 | ||
| AK171691.1 | - | 2 - 3 | 276 | 65 | ||
| BC010346.1 | - | 2 - 3 | 272 | 2811935 | 65 | |
| BY734818.1 | - | 2 - 3 | 327 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tspan17
Genomic Map