Assay Details
| Gene Symbol: Kif22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145588.1 | NP_663563.1 | 5 - 6 | 767 | 61 | |
| GenBank mRNA | AF013119.1 | - | 2 - 3 | 370 | 61 | |
| AK044826.1 | - | 2 - 3 | 2516 | 61 | ||
| AK075922.1 | - | 5 - 6 | 758 | 61 | ||
| AK084310.1 | - | 5 - 6 | 755 | 61 | ||
| AK088033.1 | - | 2 - 3 | 343 | 61 | ||
| BC003427.1 | - | 5 - 6 | 767 | 2615715 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kif22
Genomic Map