Assay Details
| Gene Symbol: Fbxl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199632.1 | NP_001186561.1 | 18 - 19 | 2040 | 79 | |
| NM_177076.4 | NP_796050.2 | 17 - 18 | 1941 | 79 | ||
| GenBank mRNA | AK029566.1 | - | 17 - 18 | 1943 | 79 | |
| AK030172.1 | - | 18 - 19 | 2024 | 79 | ||
| BC145291.1 | - | 18 - 19 | 1966 | 9053845 | 79 | |
| BC145292.1 | - | 17 - 18 | 1868 | 9053846 | 79 | |
| CO797010.1 | - | 1 - 2 | 60 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl13
Genomic Map