Assay Details
| Gene Symbol: Sidt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289668.1 | NP_001276597.1 | 23 - 24 | 2510 | 61 | |
| NM_172257.4 | NP_758461.1 | 22 - 23 | 2447 | 61 | ||
| GenBank mRNA | AK155875.1 | - | 22 - 23 | 2351 | 61 | |
| AK205943.1 | - | 1 - 2 | 43 | 61 | ||
| BC006873.1 | - | 9 - 10 | 784 | 61 | ||
| BC023957.1 | - | 22 - 23 | 2451 | 5321190 | 61 | |
| BC051101.1 | - | 23 - 24 | 2238 | 5052849 | 61 | |
| BQ749207.1 | - | 5 - 6 | 428 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sidt2
Genomic Map