Assay Details
| Gene Symbol: Sidt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289668.1 | NP_001276597.1 | 2 - 3 | 652 | 76 | |
| NM_172257.4 | NP_758461.1 | 2 - 3 | 652 | 76 | ||
| GenBank mRNA | AK081177.1 | - | 2 - 3 | 638 | 76 | |
| AK152371.1 | - | 2 - 3 | 585 | 76 | ||
| AK155875.1 | - | 2 - 3 | 556 | 76 | ||
| AK196459.1 | - | 2 - 3 | 184 | 76 | ||
| BC023957.1 | - | 2 - 3 | 656 | 5321190 | 76 | |
| BC051101.1 | - | 2 - 3 | 380 | 5052849 | 76 | |
More Information
Set Membership: |
|

Sidt2
Genomic Map