Assay Details
| Gene Symbol: Pafah1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008775.3 | NP_032801.2 | 4 - 5 | 457 | 83 | |
| GenBank mRNA | AK012164.1 | - | 4 - 5 | 392 | 83 | |
| AK153424.1 | - | 4 - 5 | 428 | 83 | ||
| BC002037.1 | - | 4 - 5 | 405 | 3486003 | 83 | |
| BC056211.2 | - | 4 - 5 | 427 | 30012448 | 83 | |
| U57747.1 | - | 4 - 5 | 389 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pafah1b2
Genomic Map