Assay Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011030.2 | NP_035160.1 | 1 - 2 | 180 | 87 | |
| GenBank mRNA | AK030574.1 | - | 1 - 2 | 112 | 87 | |
| AK149127.1 | - | 1 - 2 | 72 | 87 | ||
| AK160798.1 | - | 1 - 2 | 109 | 87 | ||
| AK161212.1 | - | 1 - 2 | 113 | 87 | ||
| AK165468.1 | - | 1 - 2 | 109 | 87 | ||
| AK169726.1 | - | 1 - 2 | 113 | 87 | ||
| BC009654.1 | - | 1 - 2 | 103 | 3484815 | 87 | |
| BE985798.1 | - | 1 - 2 | 180 | 87 | ||
| BY721363.1 | - | 1 - 2 | 161 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149127.1, AK160798.1, AK161212.1, AK165468.1, AK169726.1, BC009654.1, NM_011030.2 transcripts.
Set Membership: |
|

P4ha1
Genomic Map