Assay Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011030.2 | NP_035160.1 | 7 - 8 | 1114 | 87 | |
| GenBank mRNA | AK030574.1 | - | 7 - 8 | 1044 | 87 | |
| AK045008.1 | - | 7 - 8 | 1043 | 87 | ||
| AK149127.1 | - | 7 - 8 | 1006 | 87 | ||
| AK160798.1 | - | 7 - 8 | 1043 | 87 | ||
| AK161212.1 | - | 6 - 7 | 807 | 87 | ||
| AK165468.1 | - | 7 - 8 | 1043 | 87 | ||
| AK169726.1 | - | 7 - 8 | 1047 | 87 | ||
| BC009654.1 | - | 7 - 8 | 1037 | 3484815 | 87 | |
| U16162.1 | - | 6 - 7 | 884 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P4ha1
Genomic Map