Assay Details
| Gene Symbol: Polrmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172551.3 | NP_766139.2 | 15 - 16 | 3210 | 76 | |
| GenBank mRNA | AK003792.2 | - | 6 - 7 | 948 | 76 | |
| AK030039.1 | - | 14 - 15 | 2992 | 76 | ||
| AK042071.1 | - | 13 - 14 | 2767 | 76 | ||
| AK053356.1 | - | 15 - 16 | 3201 | 76 | ||
| AK085666.1 | - | 15 - 16 | 3203 | 76 | ||
| AK154181.1 | - | 14 - 15 | 3247 | 76 | ||
| AK154729.1 | - | 15 - 16 | 3192 | 76 | ||
| BC110697.1 | - | 15 - 16 | 3186 | 30356222 | 76 | |
| BC117940.1 | - | 15 - 16 | 3203 | 40097816 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polrmt
Genomic Map