Assay Details
| Gene Symbol: Ddx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019553.2 | NP_062426.2 | 4 - 5 | 1090 | 70 | |
| GenBank mRNA | AF159131.3 | - | 4 - 5 | 1051 | 70 | |
| AK010281.1 | - | 4 - 5 | 970 | 70 | ||
| AK145578.1 | - | 4 - 5 | 1074 | 70 | ||
| AK160095.1 | - | 4 - 5 | 1064 | 70 | ||
| BC030895.1 | - | 3 - 4 | 522 | 70 | ||
| BC043655.1 | - | 4 - 5 | 1062 | 5363948 | 70 | |
| BC059237.1 | - | 4 - 5 | 1050 | 6412782 | 70 | |
| BC060220.1 | - | 4 - 5 | 1042 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx21
Genomic Map