Assay Details
| Gene Symbol: Ccar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026201.3 | NP_080477.1 | 3 - 4 | 438 | 55 | |
| GenBank mRNA | AF465615.1 | - | 3 - 4 | 329 | 55 | |
| AK012111.1 | - | 3 - 4 | 329 | 55 | ||
| BC010199.1 | - | 3 - 4 | 316 | 55 | ||
| BC034174.1 | - | 3 - 4 | 339 | 55 | ||
| BC051052.1 | - | 3 - 4 | 340 | 55 | ||
| BC060130.1 | - | 3 - 4 | 326 | 55 | ||
| BC079652.1 | - | 3 - 4 | 312 | 6844310 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccar1
Genomic Map