Assay Details
| Gene Symbol: Hnrnph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079824.1 | NP_001073293.1 | 5 - 6 | 680 | 59 | |
| GenBank mRNA | BC147496.1 | - | 3 - 4 | 337 | 59 | |
| BQ443117.1 | - | 4 - 5 | 522 | 59 | ||
| Gene Symbol: Aff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC094538.1 | - | 521 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnph3
Genomic Map