Assay Details
| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK164324.1 | - | 1 - 2 | 76 | 105 | |
| AK164651.1 | - | 1 - 2 | 77 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164324.1 transcripts.
Set Membership: |
|

Midn
Genomic Map