Assay Details
| Gene Symbol: Ndufs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029272.3 | NP_083548.1 | 4 - 5 | 286 | 68 | |
| GenBank mRNA | AK003132.1 | - | 4 - 5 | 287 | 68 | |
| BC013503.1 | - | 4 - 5 | 263 | 4223916 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs7
Genomic Map