Assay Details
| Gene Symbol: Ndufs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029272.3 | NP_083548.1 | 7 - 8 | 603 | 76 | |
| GenBank mRNA | AK003132.1 | - | 7 - 8 | 604 | 76 | |
| BC013503.1 | - | 7 - 8 | 580 | 4223916 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
mitochondrial respiratory chain complex I assembly
oxidation-reduction process
protease binding
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity
oxidoreductase activity
metal ion binding
quinone binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
transport
mitochondrial respiratory chain complex I assembly
oxidation-reduction process
protease binding
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity
oxidoreductase activity
metal ion binding
quinone binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27281948 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27281951 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27281949 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27281950 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974273 | BC013503.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974273 | NM_029272.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24932 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140203 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974273 | BC013503.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974273 | NM_029272.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P364671 | AK003132.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27281922 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140203 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02168660 | - | - |
| Whole Mouse Genome G4122A | A_51_P364671 | AK003132.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114284 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140203 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08501907 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08501908 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08501917 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08530793 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08753741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27281949 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27281948 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27281951 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27281950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27281988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08649545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08501912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08530793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08517980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08261374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08847520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08519952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08519956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08649533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08649544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08501909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08261379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08596592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27281994 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10364750 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364745 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364749 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364751 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364746 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364748 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364747 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451312_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424313_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10364744 | - | - |
| Mouse Genome 430 2.0 Array | 1451312_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424313_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451312_at | - | - |
| Mouse Genome 430 2.0 Array | 1424313_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75406_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75406_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000006422.mm.1 | NM_029272.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006422.mm.1 | NM_029272.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006422.mm.1 | TC1000000847.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424313_PM_a_at | - | - |
| Mouse Expression Set 430 | 1451312_at | - | - |
| Mouse Expression Set 430 | 1424313_a_at | - | - |
| Mouse Expression Set 430 | 1451312_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424313_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451312_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451312_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424313_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424313_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451312_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424313_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451312_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 628243 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1740142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740142 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2688763 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2688763 | - | - |
| mouse-6 v1.1 (Illumina) | scl38688.8.1_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740142 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38688.8.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2688763 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688763 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38688.8.1_12-S | - | - |

Ndufs7
Genomic Map