Assay Details
| Gene Symbol: Med16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163276.1 | NP_001156748.1 | 13 - 14 | 2428 | 128 | |
| NM_198107.2 | NP_932775.2 | 13 - 14 | 2431 | 128 | ||
| GenBank mRNA | AK042340.1 | - | 13 - 14 | 2393 | 128 | |
| AK149345.1 | - | 13 - 14 | 2412 | 128 | ||
| BC057056.1 | - | 13 - 14 | 2385 | 6813010 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med16
Genomic Map