Assay Details
| Gene Symbol: Snx14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172926.3 | NP_766514.2 | 21 - 22 | 2267 | 157 | |
| GenBank mRNA | AK049371.1 | - | 21 - 22 | 2246 | 157 | |
| AK082389.1 | - | 21 - 22 | 2188 | 157 | ||
| AK197468.1 | - | 2 - 3 | 139 | 157 | ||
| BC043328.1 | - | 6 - 7 | 925 | 157 | ||
| BC080799.1 | - | 21 - 22 | 2252 | 4948538 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx14
Genomic Map