Assay Details
| Gene Symbol: Pgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163746.1 | NP_001157218.1 | 4 - 5 | 573 | 69 | |
| NM_028352.4 | NP_082628.3 | 4 - 5 | 573 | 69 | ||
| GenBank mRNA | AK013402.1 | - | 4 - 5 | 556 | 69 | |
| AK028066.1 | - | 4 - 5 | 563 | 69 | ||
| AK032776.1 | - | 4 - 5 | 525 | 69 | ||
| AK036561.1 | - | 4 - 5 | 559 | 69 | ||
| AK043547.1 | - | 4 - 5 | 574 | 69 | ||
| AK049337.1 | - | 4 - 5 | 525 | 69 | ||
| AK049758.1 | - | 4 - 5 | 560 | 69 | ||
| AK051706.1 | - | 4 - 5 | 513 | 69 | ||
| AK153501.1 | - | 3 - 4 | 393 | 69 | ||
| AK160913.1 | - | 4 - 5 | 574 | 69 | ||
| AK165513.1 | - | 4 - 5 | 558 | 69 | ||
| AK169082.1 | - | 4 - 5 | 538 | 69 | ||
| AK169141.1 | - | 4 - 5 | 532 | 69 | ||
| AK169787.1 | - | 4 - 5 | 547 | 69 | ||
| BC138700.1 | - | 4 - 5 | 498 | 8861722 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgm3
Genomic Map