Assay Details
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF072248.1 | - | 1 - 2 | 359 | 58 | |
| CX208033.1 | - | 1 - 2 | 251 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF072248.1 | - | 1 - 2 | 359 | 58 | |
| CX208033.1 | - | 1 - 2 | 251 | 58 | ||
Set Membership: |
|