Assay Details
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF072248.1 | - | 3 - 4 | 657 | 73 | |
| BC038264.1 | - | 3 - 4 | 458 | 5250237 | 73 | |
| BQ042794.1 | - | 2 - 3 | 225 | 73 | ||
| CK625083.1 | - | 3 - 4 | 477 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbd3
Genomic Map