Assay Details
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF072248.1 | - | 4 - 5 | 747 | 93 | |
| BC038264.1 | - | 4 - 5 | 548 | 5250237 | 93 | |
| BQ042794.1 | - | 3 - 4 | 315 | 93 | ||
| CK625083.1 | - | 4 - 5 | 567 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbd3
Genomic Map