Assay Details
| Gene Symbol: Trp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127259.1 | NP_001120731.1 | 8 - 9 | 1652 | 61 | |
| NM_001127260.1 | NP_001120732.1 | 8 - 9 | 1652 | 61 | ||
| NM_001127262.1 | NP_001120734.1 | 6 - 7 | 989 | 61 | ||
| NM_001127263.1 | NP_001120735.1 | 6 - 7 | 989 | 61 | ||
| NM_011641.2 | NP_035771.1 | 6 - 7 | 989 | 61 | ||
| GenBank mRNA | AB010152.1 | - | 6 - 7 | 963 | 61 | |
| AF075435.1 | - | 8 - 9 | 1127 | 61 | ||
| AF075436.1 | - | 8 - 9 | 1127 | 61 | ||
| AF075438.1 | - | 6 - 7 | 845 | 61 | ||
| AF075439.1 | - | 6 - 7 | 845 | 61 | ||
| BC092537.1 | - | 6 - 7 | 989 | 30504012 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp63
Genomic Map