Assay Details
| Gene Symbol: Trp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127259.1 | NP_001120731.1 | 11 - 12 | 2028 | 86 | |
| NM_001127260.1 | NP_001120732.1 | 11 - 12 | 2028 | 86 | ||
| NM_001127262.1 | NP_001120734.1 | 9 - 10 | 1365 | 86 | ||
| NM_001127264.1 | NP_001120736.1 | 9 - 10 | 1353 | 86 | ||
| NM_011641.2 | NP_035771.1 | 9 - 10 | 1365 | 86 | ||
| GenBank mRNA | AB010152.1 | - | 9 - 10 | 1339 | 86 | |
| AF075435.1 | - | 11 - 12 | 1503 | 86 | ||
| AF075436.1 | - | 11 - 12 | 1503 | 86 | ||
| AF075438.1 | - | 9 - 10 | 1221 | 86 | ||
| AF075439.1 | - | 9 - 10 | 1221 | 86 | ||
| AK137253.1 | - | 9 - 10 | 1354 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp63
Genomic Map