Assay Details
| Gene Symbol: Tbccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081368.1 | NP_001074837.1 | 5 - 6 | 1316 | 92 | |
| GenBank mRNA | AK039905.1 | - | 5 - 6 | 1141 | 92 | |
| AK040787.1 | - | 6 - 7 | 1369 | 92 | ||
| AK133804.1 | - | 5 - 6 | 1257 | 92 | ||
| AK207313.1 | - | 1 - 2 | 127 | 92 | ||
| BC082558.1 | - | 5 - 6 | 1285 | 30605020 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbccd1
Genomic Map