Assay Details
| Gene Symbol: Tbccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081368.1 | NP_001074837.1 | 6 - 7 | 1807 | 97 | |
| GenBank mRNA | AK039905.1 | - | 6 - 7 | 1632 | 97 | |
| AK040787.1 | - | 7 - 8 | 1860 | 97 | ||
| AK133804.1 | - | 6 - 7 | 1748 | 97 | ||
| AK211897.1 | - | 1 - 2 | 340 | 97 | ||
| BC079629.1 | - | 1 - 2 | 414 | 97 | ||
| BC082558.1 | - | 6 - 7 | 1776 | 30605020 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbccd1
Genomic Map