Assay Details
| Gene Symbol: Atp8b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026094.3 | NP_080370.2 | 17 - 18 | 1896 | 106 | |
| GenBank mRNA | AY364445.1 | - | 17 - 18 | 1896 | 106 | |
| BC118977.1 | - | 17 - 18 | 1896 | 40098417 | 106 | |
| BC118978.1 | - | 17 - 18 | 1896 | 40098419 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp8b3
Genomic Map