Assay Details
| Gene Symbol: Ubxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177633.4 | NP_808301.3 | 10 - 11 | 1343 | 123 | |
| GenBank mRNA | AK129218.1 | - | 6 - 7 | 941 | 123 | |
| AK147730.1 | - | 10 - 11 | 1339 | 123 | ||
| BC062904.1 | - | 9 - 10 | 1261 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2412369 | BC062904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2412369 | AK129218.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2412369 | NM_177633.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069939 | BC062904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069939 | AK129218.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069939 | AK147730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069939 | NM_177633.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373426 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151811 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937285 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 793079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968322 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917015 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303851 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21595 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12693 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178065 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148111 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113159 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147263 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2412369 | BC062904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2412369 | AK129218.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2412369 | NM_177633.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069939 | BC062904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069939 | AK129218.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069939 | AK147730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069939 | NM_177633.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147263 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148111 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113159 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08554239 | - | - |
| Whole Mouse Genome G4122A | A_52_P827677 | AK045215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298972 | BC062904.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298972 | AK129218.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298972 | AK147730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298972 | NM_177633.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1116138 | AK051319.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1116138 | NM_177633.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148111 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113159 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12775827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13052411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12991205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12791545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12891318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12904504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12894489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13023602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12662733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30651931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12664981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30652231 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10435040 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435039 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435036 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435038 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435035 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435041 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435037 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435042 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435032 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435034 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445320_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435912_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10435031 | - | - |
| Mouse Genome 430 2.0 Array | 1435912_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435912_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224111_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224111_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600004059.mm.1 | NM_177633.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004059.mm.1 | NM_177633.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004059.mm.1 | TC1600000439.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600004055.mm.1 | NM_177633.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004055.mm.1 | NM_177633.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004055.mm.1 | TC1600000439.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600004057.mm.1 | NM_177633.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004057.mm.1 | NM_177633.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004057.mm.1 | TC1600000439.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600004060.mm.1 | NM_177633.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004060.mm.1 | NM_177633.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004060.mm.1 | TC1600000439.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600004058.mm.1 | NM_177633.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004058.mm.1 | NM_177633.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004058.mm.1 | TC1600000439.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600004056.mm.1 | NM_177633.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004056.mm.1 | NM_177633.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004056.mm.1 | TC1600000439.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435912_at | - | - |
| Mouse Expression Set 430 | 1435912_PM_at | - | - |
| Mouse Expression Set 430 | 1445320_PM_at | - | - |
| Mouse Expression Set 430 | 1445320_at | - | - |
| Murine Genome U74v2 Set | 134619_PM_at | - | - |
| Murine Genome U74v2 Set | 134619_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435912_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435912_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 325689 | - | - |
| ABI Mouse Genome Survey Microarray | 633469 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5270138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270138 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223693 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223693 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2537738 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2529900 | - | - |
| mouse-6 v1.1 (Illumina) | scl00224111.2_302-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270138 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00224111.2_302-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270138 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223693 | - | - |

Ubxn7
Genomic Map