Assay Details
| Gene Symbol: Ubxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177633.4 | NP_808301.3 | 5 - 6 | 498 | 87 | |
| GenBank mRNA | AK129218.1 | - | 1 - 2 | 96 | 87 | |
| AK147730.1 | - | 5 - 6 | 494 | 87 | ||
| BC062904.1 | - | 4 - 5 | 416 | 87 | ||
| CA317987.1 | - | 4 - 5 | 416 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubxn7
Genomic Map