Assay Details
| Gene Symbol: Senp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177103.4 | NP_796077.2 | 3 - 4 | 1934 | 116 | |
| GenBank mRNA | AK028177.1 | - | 3 - 4 | 1937 | 116 | |
| AK042087.1 | - | 3 - 4 | 1827 | 116 | ||
| AK043171.1 | - | 3 - 4 | 1819 | 116 | ||
| AK136120.1 | - | 2 - 3 | 1527 | 116 | ||
| AY522918.1 | - | 3 - 4 | 1798 | 116 | ||
| BC067014.1 | - | 3 - 4 | 1701 | 6812169 | 116 | |
| BC080830.1 | - | 3 - 4 | 1818 | 30433573 | 116 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Senp5
Genomic Map