Assay Details
| Gene Symbol: Parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030253.3 | NP_084529.1 | 5 - 6 | 1795 | 134 | |
| GenBank mRNA | AK037903.1 | - | 5 - 6 | 1362 | 134 | |
| AK050032.1 | - | 5 - 6 | 1239 | 134 | ||
| BC003281.1 | - | 5 - 6 | 1237 | 3581841 | 134 | |
| BC010312.1 | - | 4 - 5 | 1220 | 134 | ||
| BC070466.1 | - | 5 - 6 | 1795 | 30615225 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp9
Genomic Map