Assay Details
| Gene Symbol: Parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030253.3 | NP_084529.1 | 6 - 7 | 2007 | 153 | |
| GenBank mRNA | AK037903.1 | - | 6 - 7 | 1574 | 153 | |
| AK050032.1 | - | 6 - 7 | 1451 | 153 | ||
| BC003281.1 | - | 6 - 7 | 1449 | 3581841 | 153 | |
| BC010312.1 | - | 5 - 6 | 1432 | 153 | ||
| BC070466.1 | - | 6 - 7 | 2007 | 30615225 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp9
Genomic Map