Assay Details
| Gene Symbol: Parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030253.3 | NP_084529.1 | 9 - 10 | 2596 | 80 | |
| GenBank mRNA | AK050032.1 | - | 9 - 10 | 2040 | 80 | |
| BC003281.1 | - | 9 - 10 | 2038 | 3581841 | 80 | |
| BC010312.1 | - | 8 - 9 | 2021 | 80 | ||
| BC070466.1 | - | 9 - 10 | 2596 | 30615225 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp9
Genomic Map