Assay Details
| Gene Symbol: Zfp280d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146224.4 | NP_666336.3 | 19 - 20 | 2413 | 84 | |
| GenBank mRNA | AK172652.1 | - | 21 - 22 | 2643 | 84 | |
| BC079878.1 | - | 19 - 20 | 2325 | 6855096 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp280d
Genomic Map