Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600014101.mm.1 |
NM_001134457.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014101.mm.1 |
NM_001134457.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014101.mm.1 |
TC1600001743.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014099.mm.1 |
NM_001134457.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014099.mm.1 |
NM_001134457.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014099.mm.1 |
TC1600001743.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014102.mm.1 |
NM_001134457.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014102.mm.1 |
NM_001134457.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014102.mm.1 |
TC1600001743.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014098.mm.1 |
NM_001134457.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014098.mm.1 |
NM_001134457.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014098.mm.1 |
TC1600001743.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014100.mm.1 |
NM_001134457.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014100.mm.1 |
NM_001134457.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014100.mm.1 |
TC1600001743.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014097.mm.1 |
NM_001134457.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014097.mm.1 |
NM_001134457.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014097.mm.1 |
TC1600001743.mm.1 |
MTA TC |