Assay Details
| Gene Symbol: Krtdap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033131.3 | NP_001028303.1 | 5 - 6 | 293 | 96 | |
| GenBank mRNA | AK131603.1 | - | 5 - 6 | 294 | 96 | |
| BC132510.1 | - | 5 - 6 | 293 | 40130787 | 96 | |
| BC132512.1 | - | 5 - 6 | 293 | 40130789 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | NM_001033131.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | AK131603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | AK131606.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | BC132510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | BC132512.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | NM_001033131.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | AK131603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | AK131606.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | BC132510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146223 | BC132512.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952897 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291550 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | NM_001033131.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | AK131603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | AK131606.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | BC132510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | BC132512.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | NM_001033131.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | AK131603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | AK131606.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | BC132510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146223 | BC132512.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01493658 | - | - |
| Whole Mouse Genome G4122A | A_52_P702887 | BC145437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | NM_001033131.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | AK131603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | AK131606.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | BC132510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | BC132512.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | NM_001033131.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | AK131603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | AK131606.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | BC132510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146223 | BC132512.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06634696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06069123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06633526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06645268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06524426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06082030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06369593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06645263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06263704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06183527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06522589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06524432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06183521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05954957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06524427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06183526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05954959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05954954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033406 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552067 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434227_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552064 | - | - |
| Mouse Genome 430 2.0 Array | 1434227_at | - | - |
| Mouse Genome 430 2.0 Array | 1434227_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64661_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64661_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700005199.mm.1 | TC0700000576.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005199.mm.1 | NM_001033131.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005199.mm.1 | NM_001033131.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005200.mm.1 | TC0700000576.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005200.mm.1 | NM_001033131.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005200.mm.1 | NM_001033131.3 | Refseq |
| Mouse Expression Set 430 | 1434227_PM_at | - | - |
| Mouse Expression Set 430 | 1434227_at | - | - |
| Murine Genome U74v2 Set | 99654_s_at | - | - |
| Murine Genome U74v2 Set | 99654_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434227_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434227_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434227_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434227_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 819086 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5220605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2868152 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2868152 | - | - |

Krtdap
Genomic Map