Assay Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183408.3 | NP_899668.1 | 2 - 3 | 710 | 83 | |
| GenBank mRNA | AF038895.1 | - | 2 - 3 | 211 | 83 | |
| AK153657.1 | - | 2 - 3 | 714 | 83 | ||
| AK160843.1 | - | 1 - 2 | 144 | 83 | ||
| BC027224.1 | - | 2 - 3 | 406 | 83 | ||
| BC139405.1 | - | 2 - 3 | 550 | 8862427 | 83 | |
| BC139408.1 | - | 2 - 3 | 550 | 8862430 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde4a
Genomic Map