Assay Details
| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172772.2 | NP_766360.2 | 2 - 3 | 960 | 119 | |
| GenBank mRNA | AK035930.1 | - | 2 - 3 | 961 | 119 | |
| AK046403.1 | - | 2 - 3 | 947 | 119 | ||
| AK046623.1 | - | 2 - 3 | 934 | 119 | ||
| AK220158.1 | - | 2 - 3 | 989 | 119 | ||
| BC049357.1 | - | 2 - 3 | 937 | 119 | ||
| BC055816.1 | - | 2 - 3 | 688 | 119 | ||
| BC058683.1 | - | 2 - 3 | 918 | 6834871 | 119 | |
| BC069845.1 | - | 2 - 3 | 673 | 119 | ||
More Information
Set Membership: |
|

Fam63b
Genomic Map