Assay Details
| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172772.2 | NP_766360.2 | 3 - 4 | 1022 | 122 | |
| GenBank mRNA | AK035930.1 | - | 3 - 4 | 1023 | 122 | |
| AK046403.1 | - | 3 - 4 | 1009 | 122 | ||
| AK046623.1 | - | 3 - 4 | 996 | 122 | ||
| AK220158.1 | - | 3 - 4 | 1051 | 122 | ||
| BC049357.1 | - | 3 - 4 | 999 | 122 | ||
| BC055816.1 | - | 3 - 4 | 750 | 122 | ||
| BC058683.1 | - | 3 - 4 | 980 | 6834871 | 122 | |
| BC069845.1 | - | 3 - 4 | 735 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam63b
Genomic Map