Assay Details
| Gene Symbol: Sltm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025690.3 | NP_079966.2 | 14 - 15 | 1941 | 152 | |
| NM_026337.1 | NP_080613.1 | 14 - 15 | 1979 | 152 | ||
| GenBank mRNA | AF462146.1 | - | 14 - 15 | 1941 | 152 | |
| AK045723.1 | - | 8 - 9 | 1041 | 152 | ||
| AK134756.1 | - | 5 - 6 | 620 | 152 | ||
| BC019992.1 | - | 3 - 4 | 311 | 152 | ||
| BC139202.1 | - | 14 - 15 | 1931 | 8862224 | 152 | |
| CD806976.1 | - | 2 - 3 | 182 | 152 | ||
| CN723089.1 | - | 3 - 4 | 398 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sltm
Genomic Map