Assay Details
| Gene Symbol: Sltm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025690.3 | NP_079966.2 | 15 - 16 | 2132 | 62 | |
| NM_026337.1 | NP_080613.1 | 15 - 16 | 2170 | 62 | ||
| GenBank mRNA | AF462146.1 | - | 15 - 16 | 2132 | 62 | |
| AK045723.1 | - | 9 - 10 | 1232 | 62 | ||
| BC019992.1 | - | 4 - 5 | 502 | 62 | ||
| BC139202.1 | - | 15 - 16 | 2122 | 8862224 | 62 | |
| CD806976.1 | - | 3 - 4 | 373 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sltm
Genomic Map