Assay Details
| Gene Symbol: Sltm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025690.3 | NP_079966.2 | 19 - 20 | 2875 | 92 | |
| NM_026337.1 | NP_080613.1 | 19 - 20 | 2913 | 92 | ||
| GenBank mRNA | AF462146.1 | - | 19 - 20 | 2875 | 92 | |
| AK134756.1 | - | 10 - 11 | 1554 | 92 | ||
| BC019992.1 | - | 8 - 9 | 1245 | 92 | ||
| BC139202.1 | - | 19 - 20 | 2865 | 8862224 | 92 | |
| BC145577.1 | - | 18 - 19 | 2684 | 92 | ||
| CF162411.1 | - | 3 - 4 | 471 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sltm
Genomic Map