Assay Details
| Gene Symbol: Sltm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025690.3 | NP_079966.2 | 2 - 3 | 297 | 109 | |
| NM_026337.1 | NP_080613.1 | 2 - 3 | 389 | 109 | ||
| GenBank mRNA | AF462146.1 | - | 2 - 3 | 297 | 109 | |
| AK011815.1 | - | 2 - 3 | 352 | 109 | ||
| AK017839.1 | - | 2 - 3 | 250 | 109 | ||
| AK077578.1 | - | 2 - 3 | 412 | 109 | ||
| AK162780.1 | - | 2 - 3 | 270 | 109 | ||
| BC139202.1 | - | 2 - 3 | 287 | 8862224 | 109 | |
| BC145577.1 | - | 2 - 3 | 287 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sltm
Genomic Map