Assay Details
| Gene Symbol: Sltm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025690.3 | NP_079966.2 | 9 - 10 | 1277 | 105 | |
| NM_026337.1 | NP_080613.1 | 9 - 10 | 1315 | 105 | ||
| GenBank mRNA | AF462146.1 | - | 9 - 10 | 1277 | 105 | |
| AK011815.1 | - | 9 - 10 | 1263 | 105 | ||
| AK017715.1 | - | 6 - 7 | 955 | 105 | ||
| AK017839.1 | - | 9 - 10 | 1176 | 105 | ||
| AK032003.1 | - | 1 - 2 | 1433 | 105 | ||
| AK045723.1 | - | 3 - 4 | 377 | 105 | ||
| AK077578.1 | - | 9 - 10 | 1338 | 105 | ||
| AK082294.1 | - | 6 - 7 | 2193 | 105 | ||
| BC139202.1 | - | 9 - 10 | 1267 | 8862224 | 105 | |
| BC145577.1 | - | 9 - 10 | 1267 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sltm
Genomic Map