Assay Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027184.1 | NP_081460.1 | 4 - 5 | 648 | 92 | |
| GenBank mRNA | AK010523.1 | - | 4 - 5 | 607 | 92 | |
| AK036084.1 | - | 4 - 5 | 416 | 92 | ||
| AK036978.1 | - | 4 - 5 | 608 | 92 | ||
| AK172246.1 | - | 4 - 5 | 606 | 92 | ||
| BC052463.1 | - | 4 - 5 | 648 | 6834748 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipmk
Genomic Map