Assay Details
| Gene Symbol: Tle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252401.1 | NP_001239330.1 | 10 - 11 | 1076 | 98 | |
| NM_019725.2 | NP_062699.1 | 10 - 11 | 1178 | 98 | ||
| GenBank mRNA | AF145958.2 | - | 10 - 11 | 933 | 98 | |
| AK144581.1 | - | 10 - 11 | 1077 | 98 | ||
| AK220533.1 | - | 9 - 10 | 1987 | 98 | ||
| BC051955.1 | - | 9 - 10 | 1131 | 98 | ||
| BC062961.1 | - | 9 - 10 | 1131 | 98 | ||
| BC071219.1 | - | 10 - 11 | 1080 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tle2
Genomic Map