Assay Details
| Gene Symbol: Tle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252401.1 | NP_001239330.1 | 12 - 13 | 1365 | 101 | |
| NM_019725.2 | NP_062699.1 | 12 - 13 | 1467 | 101 | ||
| GenBank mRNA | AF145958.2 | - | 12 - 13 | 1222 | 101 | |
| AK144581.1 | - | 12 - 13 | 1366 | 101 | ||
| AK220533.1 | - | 11 - 12 | 2276 | 102 | ||
| BC051955.1 | - | 11 - 12 | 1420 | 101 | ||
| BC062961.1 | - | 11 - 12 | 1420 | 101 | ||
| BC071219.1 | - | 12 - 13 | 1369 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tle2
Genomic Map