Assay Details
| Gene Symbol: Tle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252401.1 | NP_001239330.1 | 2 - 3 | 508 | 75 | |
| NM_019725.2 | NP_062699.1 | 2 - 3 | 508 | 75 | ||
| GenBank mRNA | AF145958.2 | - | 2 - 3 | 263 | 75 | |
| AK144581.1 | - | 2 - 3 | 509 | 75 | ||
| AK160999.1 | - | 2 - 3 | 536 | 75 | ||
| AK220533.1 | - | 2 - 3 | 1234 | 75 | ||
| BC051955.1 | - | 2 - 3 | 378 | 75 | ||
| BC062961.1 | - | 2 - 3 | 378 | 75 | ||
| BC071219.1 | - | 2 - 3 | 479 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tle2
Genomic Map