Assay Details
| Gene Symbol: Pip5k1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146687.2 | NP_001140159.1 | 11 - 12 | 1436 | 66 | |
| NM_001293646.1 | NP_001280575.1 | 11 - 12 | 1436 | 66 | ||
| NM_001293647.1 | NP_001280576.1 | 11 - 12 | 1449 | 66 | ||
| NM_008844.3 | NP_032870.2 | 11 - 12 | 1436 | 66 | ||
| GenBank mRNA | AB006916.1 | - | 11 - 12 | 1419 | 66 | |
| AK014403.1 | - | 11 - 12 | 1402 | 66 | ||
| AK041402.1 | - | 11 - 12 | 1422 | 66 | ||
| AK122319.2 | - | 11 - 12 | 1394 | 66 | ||
| AK154816.1 | - | 11 - 12 | 1428 | 66 | ||
| AK155371.1 | - | 11 - 12 | 1412 | 66 | ||
| AK170286.1 | - | 11 - 12 | 1428 | 66 | ||
| AK171576.1 | - | 11 - 12 | 1410 | 66 | ||
| BC019138.1 | - | 11 - 12 | 1385 | 5041431 | 66 | |
| BC020365.1 | - | 11 - 12 | 1398 | 66 | ||
| BC094665.1 | - | 11 - 12 | 1408 | 30356181 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pip5k1c
Genomic Map